siRNA Details
Gene Symbol: Gfra4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023967.1 | NP_076457.1 | 6 | 919 | ||
XM_006235041.2 | XP_006235103.1 | 4 | 2476 | |||
XM_006235042.2 | XP_006235104.1 | 5 | 2337 | |||
XM_006235043.2 | XP_006235105.1 | 5 | 2392 | |||
XM_006235044.2 | XP_006235106.1 | 3 | 2341 | |||
XM_006235045.2 | XP_006235107.1 | 6 | 1777 | |||
XM_008762197.1 | XP_008760419.1 | 5 | 1916 | |||
GenBank mRNA | AJ294475.1 | - | 6 | 864 | ||
AJ294476.1 | - | 6 | 919 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
