siRNA Details
Gene Symbol: Arl6ip5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023972.3 | NP_076462.1 | 2, 3 | 387 | ||
GenBank mRNA | AF240182.1 | - | 2, 3 | 387 | ||
BC061548.1 | - | 399 | 6922366 | |||
FQ213351.1 | - | 420 | ||||
FQ216190.1 | - | 420 | ||||
FQ216551.1 | - | 421 | ||||
FQ225681.1 | - | 420 | ||||
FQ228780.1 | - | 485 | ||||
FQ232358.1 | - | 420 |
