siRNA Details
Gene Symbol: Shank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031751.2 | NP_113939.2 | 2, 3 | 342 | ||
XM_008759414.1 | XP_008757636.1 | 1, 2 | 409 | |||
XM_008759415.1 | XP_008757637.1 | 1, 2 | 409 | |||
XM_008759416.1 | XP_008757638.1 | 1, 2 | 409 | |||
XM_008759417.1 | XP_008757639.1 | 1, 2 | 409 | |||
GenBank mRNA | AF102855.2 | - | 1, 2 | 254 | ||
AF131951.1 | - | 1, 2 | 263 | |||
AF141904.1 | - | 925 | ||||
AF159046.1 | - | 2, 3 | 342 |
