siRNA Details
Gene Symbol: Golph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023977.2 | NP_076467.2 | 2 | 388 | ||
GenBank mRNA | AF311954.1 | - | 2 | 279 | ||
BC092568.1 | - | 2 | 388 | 7307277 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025673, AK014644, AK133763, AK015543, AK139545, and others.
Gene Ontology Categories:
