siRNA Details
Gene Symbol: Trib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023985.1 | NP_076475.1 | 3 | 1318 | ||
GenBank mRNA | AF205438.1 | - | 3 | 694 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023985.1 | NP_076475.1 | 3 | 1318 | ||
GenBank mRNA | AF205438.1 | - | 3 | 694 |