siRNA Details
Gene Symbol: Birc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023987.3 | NP_076477.3 | 8 | 1652 | ||
XM_006242499.2 | XP_006242561.1 | 7 | 3615 | |||
XM_006242500.2 | XP_006242562.1 | 8 | 2385 | |||
XM_006242502.2 | XP_006242564.1 | 7 | 1884 | |||
XM_006242503.2 | XP_006242565.1 | 7 | 1843 | |||
GenBank mRNA | AF183430.1 | - | 6 | 1420 | ||
BC083555.1 | - | 8 | 1652 | 7112819 |
