siRNA Details
Gene Symbol: Birc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023987.3 | NP_076477.3 | 3, 4 | 1077 | ||
XM_006242499.2 | XP_006242561.1 | 2, 3 | 3040 | |||
XM_006242500.2 | XP_006242562.1 | 3, 4 | 1810 | |||
XM_006242502.2 | XP_006242564.1 | 2, 3 | 1309 | |||
XM_006242503.2 | XP_006242565.1 | 2, 3 | 1268 | |||
GenBank mRNA | AF183430.1 | - | 1, 2 | 845 | ||
BC083555.1 | - | 3, 4 | 1077 | 7112819 |
