siRNA Details
Gene Symbol: Birc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023987.3 | NP_076477.3 | 7, 8 | 1564 | ||
XM_006242499.2 | XP_006242561.1 | 6, 7 | 3527 | |||
XM_006242500.2 | XP_006242562.1 | 7, 8 | 2297 | |||
XM_006242502.2 | XP_006242564.1 | 6, 7 | 1796 | |||
XM_006242503.2 | XP_006242565.1 | 6, 7 | 1755 | |||
GenBank mRNA | AF183430.1 | - | 5, 6 | 1332 | ||
BC083555.1 | - | 7, 8 | 1564 | 7112819 |
