siRNA Details
Gene Symbol: Mepe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024142.1 | NP_077056.1 | 3 | 423 | ||
XM_006250634.2 | XP_006250696.1 | 2 | 491 | |||
XM_006250635.2 | XP_006250697.1 | 2 | 2057 | |||
GenBank mRNA | AF260922.1 | - | 3 | 423 | ||
AF530558.1 | - | 3 | 420 | |||
AF530559.1 | - | 2 | 399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
