siRNA Details
Gene Symbol: Slc27a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024143.2 | NP_077057.1 | 7 | 1609 | ||
GenBank mRNA | AF242189.1 | - | 7 | 1610 | ||
BC091147.1 | - | 7 | 1632 | 7370589 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
