siRNA Details
Gene Symbol: Fgfr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024146.1 | NP_077060.1 | 10 | 1520 | ||
XM_006253323.2 | XP_006253385.1 | 10 | 1522 | |||
XM_006253324.2 | XP_006253386.1 | 11 | 1735 | |||
XM_006253325.2 | XP_006253387.1 | 9 | 1261 | |||
XM_006253326.2 | XP_006253388.1 | 9 | 1255 | |||
XM_006253327.1 | XP_006253389.1 | 10 | 1536 | |||
GenBank mRNA | D12498.1 | - | 10 | 1520 | ||
S54008.1 | - | 10 | 1541 |
