siRNA Details
Gene Symbol: Fgfr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024146.1 | NP_077060.1 | 12, 13 | 1856 | ||
XM_006253323.2 | XP_006253385.1 | 12, 13 | 1858 | |||
XM_006253324.2 | XP_006253386.1 | 13, 14 | 2071 | |||
XM_006253325.2 | XP_006253387.1 | 11, 12 | 1597 | |||
XM_006253326.2 | XP_006253388.1 | 11, 12 | 1591 | |||
XM_006253327.1 | XP_006253389.1 | 12, 13 | 1872 | |||
GenBank mRNA | D12498.1 | - | 12, 13 | 1856 | ||
S54008.1 | - | 12, 13 | 1877 |
