siRNA Details
Gene Symbol: Arf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024149.1 | NP_077063.1 | 2, 3 | 245 | ||
GenBank mRNA | BC087692.1 | - | 2, 3 | 207 | 7310424 | |
L12384.1 | - | 2, 3 | 245 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
