siRNA Details
Gene Symbol: Cfi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024157.1 | NP_077071.1 | 12 | 1443 | ||
XM_008761526.1 | XP_008759748.1 | 13 | 1781 | |||
GenBank mRNA | BC089798.1 | - | 12 | 1448 | 7321559 | |
Y18965.1 | - | 12 | 1443 |
