siRNA Details
Gene Symbol: Fabp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024162.1 | NP_077076.1 | 3 | 354 | ||
GenBank mRNA | FQ217465.1 | - | 3 | 358 | ||
J02773.1 | - | 3 | 354 | |||
M18034.1 | - | 3 | 341 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
