siRNA Details
Gene Symbol: Fabp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024162.1 | NP_077076.1 | 3 | 365 | ||
GenBank mRNA | FQ217465.1 | - | 3 | 369 | ||
J02773.1 | - | 3 | 365 | |||
M18034.1 | - | 3 | 352 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fabp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024162.1 | NP_077076.1 | 3 | 365 | ||
GenBank mRNA | FQ217465.1 | - | 3 | 369 | ||
J02773.1 | - | 3 | 365 | |||
M18034.1 | - | 3 | 352 |