siRNA Details
Gene Symbol: Frk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024368.1 | NP_077344.1 | 1 | 599 | ||
XM_006256517.2 | XP_006256579.1 | 1 | 750 | |||
XM_008773006.1 | XP_008771228.1 | 1 | 750 | |||
GenBank mRNA | U09583.1 | - | 1 | 599 |
