siRNA Details
Gene Symbol: Fstl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024369.2 | NP_077345.1 | 4 | 357 | ||
GenBank mRNA | AY325203.1 | - | 456 | |||
BC087014.1 | - | 4 | 357 | 7189359 | ||
U06864.1 | - | 4 | 332 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
