siRNA Details
Gene Symbol: Hhex | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024385.1 | NP_077361.1 | 3, 4 | 597 | ||
GenBank mRNA | BC088135.1 | - | 3, 4 | 602 | 7366155 | |
D86383.1 | - | 3, 4 | 597 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
