siRNA Details
Gene Symbol: Hmgcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024386.1 | NP_077362.1 | 5 | 446 | ||
GenBank mRNA | BC061797.1 | - | 5 | 448 | 5598033 | |
FQ209392.1 | - | 5 | 444 | |||
FQ209616.1 | - | 5 | 450 | |||
FQ234382.1 | - | 5 | 444 | |||
Y10054.1 | - | 5 | 446 |

More Information
Other Species Targeted

Gene Ontology Categories:
