siRNA Details
Gene Symbol: Hpgd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024390.2 | NP_077366.2 | 4 | 384 | ||
GenBank mRNA | BC062399.1 | - | 4 | 384 | 5599288 | |
U44750.1 | - | 4 | 400 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008278, AK146038, U44389, AK002472, BC021157.
Gene Ontology Categories:
