siRNA Details
Gene Symbol: Hsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024393.1 | NP_077369.1 | 4 | 578 | ||
XM_006241888.2 | XP_006241950.1 | 4 | 567 | |||
XM_006241889.2 | XP_006241951.1 | 4 | 568 | |||
XM_006241890.2 | XP_006241952.1 | 4 | 567 | |||
XM_006241891.2 | XP_006241953.1 | 4 | 566 | |||
GenBank mRNA | X83094.1 | - | 2 | 150 |

More Information
Other Species Targeted

Gene Ontology Categories:
