siRNA Details
Gene Symbol: Abca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024396.1 | NP_077372.1 | 29 | 4777 | ||
XM_006233645.2 | XP_006233707.1 | 30 | 4886 | |||
XM_008761609.1 | XP_008759831.1 | 29 | 7453 | |||
GenBank mRNA | AB037937.1 | - | 29 | 4777 |
