siRNA Details
Gene Symbol: Aco2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024398.2 | NP_077374.2 | 10, 11 | 1303 | ||
GenBank mRNA | AJ243266.1 | - | 10, 11 | 1304 | ||
BC061999.1 | - | 10, 11 | 1303 | 5621226 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
