siRNA Details
Gene Symbol: Clcnka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053327.1 | NP_445779.1 | 16 | 1758 | ||
XM_008764291.1 | XP_008762513.1 | 16 | 1874 | |||
XM_008764292.1 | XP_008762514.1 | 16 | 1844 | |||
GenBank mRNA | BC081761.1 | - | 16 | 1824 | 7108273 | |
D13927.1 | - | 16 | 1758 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
