siRNA Details
Gene Symbol: Pdgfc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031317.1 | NP_112607.1 | 3 | 436 | ||
GenBank mRNA | AB033830.1 | - | 3 | 436 | ||
AF508348.1 | - | 2 | 314 |

More Information
Other Species Targeted

Gene Ontology Categories:
