siRNA Details
Gene Symbol: Bhlhe40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053328.1 | NP_445780.1 | 3 | 565 | ||
GenBank mRNA | AF009330.1 | - | 3 | 565 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011498, BC010720, AK090032, AF010305, AF364051, and others.
Gene Ontology Categories:
