siRNA Details
Gene Symbol: Myh10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031520.1 | NP_113708.1 | 13 | 1613 | ||
XM_006246821.2 | XP_006246883.1 | 13 | 1660 | |||
XM_008767880.1 | XP_008766102.1 | 13 | 1770 | |||
XM_008767881.1 | XP_008766103.1 | 13 | 1486 | |||
GenBank mRNA | AF139055.1 | - | 13 | 1613 |
