siRNA Details
Gene Symbol: Ica1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030844.2 | NP_110471.2 | 9 | 1076 | ||
XM_006236092.2 | XP_006236154.1 | 9 | 1172 | |||
XM_006236093.2 | XP_006236155.1 | 9 | 1394 | |||
XM_006236094.2 | XP_006236156.1 | 8 | 1022 | |||
XM_006236095.2 | XP_006236157.1 | 8 | 1008 | |||
XM_008762728.1 | XP_008760950.1 | 9 | 1172 | |||
GenBank mRNA | BC063810.1 | - | 9 | 1095 | ||
BC078812.1 | - | 9 | 1076 | 7112043 | ||
L20900.1 | - | 9 | 1282 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
