siRNA Details
Gene Symbol: Ica1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030844.2 | NP_110471.2 | 8 | 973 | ||
XM_006236092.2 | XP_006236154.1 | 8 | 1069 | |||
XM_006236093.2 | XP_006236155.1 | 8 | 1291 | |||
XM_006236094.2 | XP_006236156.1 | 7 | 919 | |||
XM_006236095.2 | XP_006236157.1 | 7 | 905 | |||
XM_008762728.1 | XP_008760950.1 | 8 | 1069 | |||
GenBank mRNA | BC063810.1 | - | 8 | 992 | ||
BC078812.1 | - | 8 | 973 | 7112043 | ||
L20900.1 | - | 8 | 1179 |

More Information
Other Species Targeted

Gene Ontology Categories:
