siRNA Details
Gene Symbol: Emp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030847.1 | NP_110474.1 | 3 | 351 | ||
XM_006229158.2 | XP_006229220.1 | 3 | 415 | |||
XM_008759418.1 | XP_008757640.1 | 3 | 293 | |||
GenBank mRNA | BC088131.1 | - | 3 | 347 | 7365466 | |
FQ233038.1 | - | 3 | 338 | |||
Y10889.2 | - | 3 | 351 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
