siRNA Details
Gene Symbol: Bmpr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030849.1 | NP_110476.1 | 2, 3 | 301 | ||
GenBank mRNA | D17667.1 | - | 1, 2 | 76 | ||
D38082.1 | - | 1, 2 | 158 | |||
S75359.1 | - | 2, 3 | 301 |
Gene Symbol: LOC686129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767400.1 | XP_008765622.1 | 1 | 312 |

More Information
Other Species Targeted

Gene Ontology Categories:
