siRNA Details
Gene Symbol: Bmpr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030849.1 | NP_110476.1 | 10, 11 | 1576 | ||
GenBank mRNA | D17667.1 | - | 9, 10 | 1351 | ||
D38082.1 | - | 9, 10 | 1433 | |||
S75359.1 | - | 10, 11 | 1576 |
Gene Symbol: LOC686129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767400.1 | XP_008765622.1 | 2 | 1596 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
