siRNA Details
Gene Symbol: Bhmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030850.1 | NP_110477.1 | 4 | 468 | ||
GenBank mRNA | AF038870.1 | - | 4 | 468 | ||
BC078811.1 | - | 4 | 455 | 7130736 | ||
U96133.1 | - | 4 | 457 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
