siRNA Details
Gene Symbol: Mdk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234591.2 | XP_006234653.1 | 5 | 734 | ||
XM_006234592.2 | XP_006234654.1 | 5 | 682 | |||
XM_006234593.2 | XP_006234655.1 | 5 | 1359 | |||
GenBank mRNA | AF315950.1 | - | 5 | 648 | ||
BC157805.1 | - | 5 | 600 | 9026085 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
