siRNA Details
Gene Symbol: Msn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030863.1 | NP_110490.1 | 13 | 1760 | ||
GenBank mRNA | AF004811.1 | - | 13 | 1760 | ||
FQ230914.1 | - | 1847 |

More Information
Other Species Targeted

Gene Ontology Categories:
