siRNA Details
Gene Symbol: Nfix | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030866.1 | NP_110493.1 | 2, 3 | 606 | ||
XM_006255307.2 | XP_006255369.1 | 2, 3 | 690 | |||
XM_006255308.2 | XP_006255370.1 | 2, 3 | 746 | |||
XM_006255310.2 | XP_006255372.1 | 2, 3 | 1591 | |||
XM_008772418.1 | XP_008770640.1 | 2, 3 | 689 | |||
XM_008772419.1 | XP_008770641.1 | 2, 3 | 687 | |||
GenBank mRNA | AB012235.1 | - | 2, 3 | 606 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001081982, NM_001081981, NM_010906, Y07689, BC090626, and others.
Gene Ontology Categories:
