siRNA Details
Gene Symbol: Nfkbib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030867.2 | NP_110494.2 | 2 | 518 | ||
GenBank mRNA | AF246634.1 | - | 2 | 341 | ||
BC085729.1 | - | 2 | 518 | 7114624 | ||
FQ228042.1 | - | 2 | 315 | |||
FQ235103.1 | - | 2 | 463 |
