siRNA Details
Gene Symbol: Pfn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030873.1 | NP_110500.1 | 238 | |||
XM_006232390.2 | XP_006232452.1 | 2 | 363 | |||
GenBank mRNA | AF228736.1 | - | 238 | |||
AF228737.1 | - | 238 | ||||
AY004289.1 | - | 238 | ||||
BC087013.1 | - | 241 |
Gene Symbol: LOC100909840 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749282.2 | XP_003749330.1 | 2 | 279 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
