siRNA Details
Gene Symbol: Sgpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_343081.7 | XP_343082.4 | 1, 2 | 787 | ||
GenBank mRNA | AF329638.1 | - | 1, 2 | 285 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sgpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_343081.7 | XP_343082.4 | 1, 2 | 787 | ||
GenBank mRNA | AF329638.1 | - | 1, 2 | 285 |