siRNA Details
Gene Symbol: Sgpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_343081.7 | XP_343082.4 | 3 | 894 | ||
GenBank mRNA | AF329638.1 | - | 3 | 392 |

More Information
Other Species Targeted

Gene Ontology Categories:
