siRNA Details
Gene Symbol: Adar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031006.1 | NP_112268.1 | 2 | 1343 | ||
XM_006232774.2 | XP_006232836.1 | 3 | 1567 | |||
XM_006232775.2 | XP_006232837.1 | 3 | 1566 | |||
XM_006232776.2 | XP_006232838.1 | 2 | 4529 | |||
XM_006232777.2 | XP_006232839.1 | 2 | 1375 | |||
XM_006232778.2 | XP_006232840.1 | 2 | 1327 | |||
GenBank mRNA | FQ212262.1 | - | 1551 | |||
FQ212270.1 | - | 1551 | ||||
U18942.1 | - | 2 | 1343 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001146296, NM_019655, NM_001038587, AF291876, CT010323, and others.
Gene Ontology Categories:
