siRNA Details
Gene Symbol: Adar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031006.1 | NP_112268.1 | 2 | 1256 | ||
XM_006232774.2 | XP_006232836.1 | 3 | 1480 | |||
XM_006232775.2 | XP_006232837.1 | 3 | 1479 | |||
XM_006232776.2 | XP_006232838.1 | 2 | 4442 | |||
XM_006232777.2 | XP_006232839.1 | 2 | 1288 | |||
XM_006232778.2 | XP_006232840.1 | 2 | 1240 | |||
GenBank mRNA | FQ212262.1 | - | 1464 | |||
FQ212270.1 | - | 1464 | ||||
U18942.1 | - | 2 | 1256 |

More Information
Other Species Targeted

Gene Ontology Categories:
