siRNA Details
Gene Symbol: Anpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031012.1 | NP_112274.1 | 10 | 1668 | ||
XM_006229407.2 | XP_006229469.1 | 11 | 2088 | |||
XM_006229408.2 | XP_006229470.1 | 11 | 2109 | |||
XM_006229409.1 | XP_006229471.1 | 11 | 2145 | |||
GenBank mRNA | M25073.1 | - | 10 | 1668 | ||
M26710.1 | - | 10 | 1715 | |||
Y18765.1 | - | 10 | 1058 |
