siRNA Details
Gene Symbol: Mapk14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031020.2 | NP_112282.2 | 11 | 1301 | ||
XM_006256142.2 | XP_006256204.1 | 11 | 1013 | |||
GenBank mRNA | AB000783.1 | - | 11 | 1300 | ||
AF346293.1 | - | 11 | 998 | |||
BC092193.1 | - | 11 | 1301 | 7313217 | ||
U91847.1 | - | 11 | 998 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
