siRNA Details
Gene Symbol: Dync1li2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031026.1 | NP_112288.1 | 12 | 1444 | ||
XM_006255053.2 | XP_006255115.1 | 13 | 1643 | |||
XM_006255054.2 | XP_006255116.1 | 12 | 1601 | |||
GenBank mRNA | AB008521.1 | - | 8 | 695 | ||
AB008522.1 | - | 7 | 710 | |||
AB008523.1 | - | 7 | 653 | |||
AB008524.1 | - | 8 | 752 | |||
BC089794.1 | - | 13 | 1450 | 7115029 | ||
U15138.1 | - | 12 | 1444 |
