siRNA Details
Gene Symbol: Gabrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031029.1 | NP_112291.1 | 9 | 1285 | ||
XM_008767594.1 | XP_008765816.1 | 10 | 1871 | |||
XM_008767595.1 | XP_008765817.1 | 10 | 1877 | |||
GenBank mRNA | FQ222085.1 | - | 10 | 1452 | ||
U95368.1 | - | 9 | 1285 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
