siRNA Details
Gene Symbol: Gnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031037.2 | NP_112299.1 | 7 | 892 | ||
GenBank mRNA | AF277892.1 | - | 6 | 673 | ||
AF397193.1 | - | 3 | 472 | |||
BC065579.1 | - | 7 | 892 | 5598923 | ||
DQ120485.1 | - | 6 | 657 | |||
DQ120486.1 | - | 6 | 657 | |||
FQ209560.1 | - | 3 | 280 | |||
U34959.1 | - | 6 | 664 |

More Information
Other Species Targeted

Gene Ontology Categories:
