siRNA Details
Gene Symbol: Gtf2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031041.1 | NP_112303.1 | 4 | 404 | ||
GenBank mRNA | BC085345.1 | - | 4 | 409 | 7303747 | |
FQ229769.1 | - | 4 | 392 | |||
FQ231337.1 | - | 4 | 434 | |||
X65948.1 | - | 4 | 404 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
