siRNA Details
Gene Symbol: Aldh6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031057.2 | NP_112319.2 | 6 | 705 | ||
XM_006240354.2 | XP_006240416.1 | 6 | 1070 | |||
XM_008764862.1 | XP_008763084.1 | 5 | 609 | |||
XM_008764863.1 | XP_008763085.1 | 6 | 707 | |||
GenBank mRNA | DN948946.1 | - | 6 | 702 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
